N-Carbobenzoxy-D-methionine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-Benzyloxycarbonyl-D-methionine, Cbz-D-Met-OH, Z-D-Met-OH, N-Cbz-D-methionine

C0665-5G C0665-100MG C0665-25G
TCC0665-5GEA 64.46 CAD
TCC0665-5G TCC0665-100MG TCC0665-25G
N-Carbobenzoxy-D-methionine ≥98.0% (by titrimetric analysis)
N-Carbobenzoxy-D-methionine

CAS Number: 28862-80-8
MDL Number: MFCD00026043
Molecular Formula: C13H17NO4S
Molecular Weight: 283.34
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 69
Specific rotation [a]20/D: 26 deg (C=1, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-D-methionine, Z-D-Met-OH

Formula: C₁₃H₁₇NO₄S
MW: 283.34 g/mol
Melting Pt: 69 °C
MDL Number: MFCD00026043
CAS Number: 28862-80-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6d32f27e42595faf8a129253dc07137dc589953&vid=d6d32f27e42595faf8a129253dc07137dc589953&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR