N-Carbobenzoxy-D-glutamic acid ≥98.0%
Supplier: TCI America
Synonyms:
N-Benzyloxycarbonyl-D-glutamic acid, N-Cbz-D-glutamic Acid, Z-D-Glu-OH, Cbz-D-Glu-OH
C0663-25G
C0663-5G
TCC0663-100MGEA
26.04
CAD
TCC0663-100MG
TCC0663-25G
TCC0663-5G
N-Carbobenzoxy-D-glutamic acid ≥98.0%
N-Carbobenzoxy-D-glutamic acid
CAS Number: 63648-73-7
MDL Number: MFCD00063193
Molecular Formula: C13H15NO6
Molecular Weight: 281.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 120
Specific rotation [a]20/D: 7.5 deg (C=8, AcOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-D-glutamic Acid, Z-D-Glu-OH
Formula:
C₁₃H₁₅NO₆ MW: 281.26 g/mol Melting Pt: 120 °C |
MDL Number:
MFCD00063193 CAS Number: 63648-73-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...