Nɑ-Carbobenzoxy-DL-asparagine ≥99.0% (by titrimetric analysis)

Supplier: TCI America

C0661-1G C0661-10G
TCC0661-001GEA 26.86 CAD
TCC0661-001G TCC0661-010G
Nɑ-Carbobenzoxy-DL-asparagine ≥99.0% (by titrimetric analysis)
Nalpha-Carbobenzoxy-DL-asparagine

CAS Number: 29880-22-6
MDL Number: MFCD00065101
Molecular Formula: C12H14N2O5
Molecular Weight: 266.25
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 165


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Cbz-DL-asparagine, Z-DL-Asn-OH

Formula: C₁₂H₁₄N₂O₅
Melting Pt: 165 °C
MDL Number: MFCD00065101
CAS Number: 29880-22-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5ea86913b26d9e45ca431ebb8f3c17cb4cbe121&vid=c5ea86913b26d9e45ca431ebb8f3c17cb4cbe121&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR