N-Carbobenzoxy-DL-phenylalanine ≥98.0%

Supplier: TCI America

Synonyms: Z-DL-Phe-OH, N-Cbz-DL-phenylalanine, Cbz-DL-Phe-OH, ((Benzyloxy)carbonyl)phenylalanine

C0643-25G C0643-10G
TCC0643-025GEA 126.4 CAD
TCC0643-025G TCC0643-010G
N-Carbobenzoxy-DL-phenylalanine ≥98.0%
N-Carbobenzoxy-DL-phenylalanine

CAS Number: 3588-57-6
MDL Number: MFCD00063150
Molecular Formula: C17H17NO4
Molecular Weight: 299.33
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 104


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-DL-phenylalanine, Z-DL-Phe-OH

Formula: C₁₇H₁₇NO₄
MW: 299.33 g/mol
Melting Pt: 104 °C
MDL Number: MFCD00063150
CAS Number: 3588-57-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed0437ac52fa4dd994622d5920fe88c92b6b6bd0&vid=ed0437ac52fa4dd994622d5920fe88c92b6b6bd0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR