N-Carbobenzoxy-DL-leucine ≥98.0%

Supplier: TCI America

Synonyms: N-Cbz-DL-leucine, Z-DL-Leu-OH

C0642-5G C0642-1G
TCC0642-5GEA 254.38 CAD
TCC0642-5G TCC0642-001G
N-Carbobenzoxy-DL-leucine ≥98.0%
N-Carbobenzoxy-DL-leucine

CAS Number: 3588-60-1
MDL Number: MFCD00065129
Molecular Formula: C14H19NO4
Molecular Weight: 265.31
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 54


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-DL-leucine, Z-DL-Leu-OH

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
Melting Pt: 54 °C
MDL Number: MFCD00065129
CAS Number: 3588-60-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16c477561cfa7d33c995c7515c714107cae7b932&vid=16c477561cfa7d33c995c7515c714107cae7b932&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR