N-Carbobenzoxy-β-alanine ≥98.0%

Supplier: TCI America

Synonyms: Carbobenzyloxy-beta-alanine

C0640-10G C0640-1G
TCC0640-010GEA 172.22 CAD
TCC0640-010G TCC0640-001G
N-Carbobenzoxy-β-alanine ≥98.0%
N-Carbobenzoxy-beta-alanine

CAS Number: 2304-94-1
MDL Number: MFCD00037292
Molecular Formula: C11H13NO4
Molecular Weight: 223.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 105


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-beta-alanine, Z-beta-Ala-OH

Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
Melting Pt: 105 °C
MDL Number: MFCD00037292
CAS Number: 2304-94-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=503b5ff7f824003d029edbbbd129ea872c7befba&vid=503b5ff7f824003d029edbbbd129ea872c7befba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR