2-Amino-4,6-dimethylpyridine ≥98.0%
Supplier: TCI America
A0731-5G
A0731-25G
TCA0731-5GEA
74.26
CAD
TCA0731-5G
TCA0731-025G
2-Amino-4,6-dimethylpyridine ≥98.0%
2-Amino-4,6-dimethylpyridine
CAS Number: 5407-87-4
MDL Number: MFCD00006322
Molecular Formula: C7H10N2
Molecular Weight: 122.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 231
Melting point (°C): 67
Flash Point (°C): 121
Other Product Names / Synonyms: 2-Amino-4,6-dimethylpyridine
Formula:
C₇H₁₀N₂ MW: 122.1700 g/mol Boiling Pt: 231 °C Melting Pt: 67 °C Flash Pt: 121 °C |
MDL Number:
MFCD00006322 CAS Number: 5407-87-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...