N-Carbobenzoxy-DL-valine ≥99.0% (by titrimetric analysis)

Supplier: TCI America

C0634-1G C0634-10G
TCC0634-001GEA 57.83 CAD
TCC0634-001G TCC0634-010G
N-Carbobenzoxy-DL-valine ≥99.0% (by titrimetric analysis)
N-Carbobenzoxy-DL-valine

CAS Number: 3588-63-4
MDL Number: MFCD00065127
Molecular Formula: C13H17NO4
Molecular Weight: 251.28
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 78


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-DL-valine, Z-DL-Val-OH

Formula: C₁₃H₁₇NO₄
Melting Pt: 78 °C
MDL Number: MFCD00065127
CAS Number: 3588-63-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e345a7c1f9f9ca259291740fa6eb43c87f6cf80f&vid=e345a7c1f9f9ca259291740fa6eb43c87f6cf80f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR