N-Carbobenzoxy-DL-methionine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
C0630-10G
C0630-1G
TCC0630-010GEA
116.92
CAD
TCC0630-010G
TCC0630-001G
N-Carbobenzoxy-DL-methionine ≥98.0% (by HPLC, titration analysis)
N-Carbobenzoxy-DL-methionine
CAS Number: 4434-61-1
MDL Number: MFCD00065123
Molecular Formula: C13H17NO4S
Molecular Weight: 283.34
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 113
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-DL-methionine, Z-DL-Met-OH
Formula:
C₁₃H₁₇NO₄S Melting Pt: 113 °C |
MDL Number:
MFCD00065123 CAS Number: 4434-61-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...