Oil of cedar wood

Supplier: TCI America
Warning

C0626-500ML C0626-25ML C0626-100ML
TCC0626-500MLEA 281.24 CAD
TCC0626-500ML TCC0626-025ML TCC0626-100ML
Oil of cedar wood
Oil of cedar wood

CAS Number: 8000-27-9
MDL Number: MFCD00132766
Form: Clear Liquid
Color: Yellow
Flash Point (°C): 113
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: Cedarwood Oil

Density: 0.95 (20 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00146327
CAS Number: 8000-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR