Isopropyl carbamate ≥98.0% (by GC, total nitrogen)
Supplier: TCI America
Synonyms:
Carbamic Acid Isopropyl Ester
C0614-250G
C0614-25G
TCC0614-250GEA
1041.38
CAD
TCC0614-250G
TCC0614-025G
Isopropyl carbamate ≥98.0% (by GC, total nitrogen)
Isopropyl carbamate
CAS Number: 1746-77-6
MDL Number: MFCD00025463
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Boiling point (°C): 183
Melting point (°C): 94
Other Product Names / Synonyms: Carbamic Acid Isopropyl Ester
Formula:
C₄H₉NO₂ MW: 103.12 g/mol Boiling Pt: 183 °C Melting Pt: 94 °C |
MDL Number:
MFCD00025463 CAS Number: 1746-77-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...