β-Carotene ≥97.0% (by UV method)

Supplier: TCI America

C0560-1G C0560-5G
TCC0560-001GEA 80.82 CAD
TCC0560-001G TCC0560-005G
β-Carotene ≥97.0% (by UV method)
beta-Carotene

CAS Number: 7235-40-7
MDL Number: MFCD00001556
Molecular Formula: C40H56
Molecular Weight: 536.89
Purity/Analysis Method: >97.0% (E)
Form: Crystal
Melting point (°C): 184
Flash Point (°C): 222


Product Categories: Bioscience, Antioxidants, Vitamins, Terpenes (Others)
Other Product Names / Synonyms: Provitamin A

Formula: C₄₀H₅₆
Melting Pt: 184 °C
Flash Pt: 222 °C
MDL Number: MFCD00001556
CAS Number: 7235-40-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=520a6ba995f94ba2e70ccbbf10ef43de2c2ef049&vid=520a6ba995f94ba2e70ccbbf10ef43de2c2ef049&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (E)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR