Choline hydrogen tartrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C0553-25G C0553-500G
TCC0553-025GEA 34.76 CAD
TCC0553-025G TCC0553-500G
Choline hydrogen tartrate ≥98.0% (by titrimetric analysis)
Choline hydrogen tartrate

CAS Number: 87-67-2
MDL Number: MFCD00036332
Molecular Formula: C9H19NO7
Molecular Weight: 253.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 150
Specific rotation [a]20/D: 17 deg (C=10, H2O)


Product Categories: Bioscience, Vitamin Related Compounds
Other Product Names / Synonyms: Choline Hydrogen L-(+)-Tartrate

Formula: C₉H₁₉NO₇
Melting Pt: 150 °C
MDL Number: MFCD00036332
CAS Number: 87-67-2
EINECS: 201-763-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcf4f82c5f57690ad194f7eecf6fb79fac634fad&vid=fcf4f82c5f57690ad194f7eecf6fb79fac634fad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR