trans-Cinnamamide ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

C0536-25G C0536-5G
TCC0536-025GEA 206.98 CAD
TCC0536-025G TCC0536-5G
trans-Cinnamamide ≥98.0% (by GC, total nitrogen)
trans-Cinnamamide

CAS Number: 22031-64-7
MDL Number: MFCD00008033
Molecular Formula: C9H9NO
Molecular Weight: 147.18
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 150


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: trans-3-Phenylacrylamide

Formula: C₉H₉NO
Melting Pt: 150 °C
MDL Number: MFCD00008033
CAS Number: 22031-64-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c922ea5c06da94bd2a2009f877c3fd94ab1ff0bb&vid=c922ea5c06da94bd2a2009f877c3fd94ab1ff0bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR