Cytidine hemisulfate salt ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C0525-1G C0525-100MG
TCC0525-001GEA 96.38 CAD
TCC0525-001G TCC0525-100MG
Cytidine hemisulfate salt ≥98.0% (by HPLC, titration analysis)
Cytidine hemisulfate salt

CAS Number: 32747-18-5
MDL Number: MFCD00151091
Molecular Formula: C9H13N3O5
Molecular Weight: 584.51
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 225
Specific rotation [a]20/D: 36 deg (C=2, H2O)

Formula: C18H26N6O10·H2SO4
Melting Pt: 225 °C
MDL Number: MFCD00151091
CAS Number: 32747-18-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54de11ccf2846a33bb45f88b94a9755412a6074a&vid=54de11ccf2846a33bb45f88b94a9755412a6074a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR