Cytidine-5'-monophosphate disodium salt hydrate ≥99.0% (by HPLC)

Supplier: TCI America

C0524-5G C0524-1G
TCC0524-5GEA 45.82 CAD
TCC0524-5G TCC0524-001G
Cytidine-5'-monophosphate disodium salt hydrate ≥99.0% (by HPLC)
Cytidine-5'-monophosphate disodium salt hydrate

CAS Number: 6757-06-8
MDL Number: MFCD00064355
Molecular Formula: C9H14N3O8P
Molecular Weight: 367.16
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 8 deg (C=2, H2O)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Stains & Dyes, Nucleotides and Their Analogs
Other Product Names / Synonyms: 5'-Cytidylic Acid Disodium Salt, 5'-CMP 2Na

Formula: C₉H₁₄N₃O₈P
MDL Number: MFCD00064355
CAS Number: 6757-06-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=386ddd47e38e0e44fd839ed2e6526271f614eab9&vid=583565fe49920593a19c43b0b172812538feb049&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR