Cytidine-5'-diphosphate trisodium salt hydrate ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: CDP trisodium salt, 5'-CDP 3Na

C0523-1G C0523-100MG C0523-10MG
TCC0523-1GEA 423.44 CAD
TCC0523-1G TCC0523-100MG TCC0523-10MG
Cytidine-5'-diphosphate trisodium salt hydrate ≥95.0% (by HPLC, titration analysis)
Cytidine-5'-diphosphate trisodium salt hydrate

CAS Number: 34393-59-4
MDL Number: MFCD00069770
Molecular Formula: C9H15N3O11P2
Molecular Weight: 469.12
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Bioscience, Nucleotides and Their Analogs
Other Product Names / Synonyms: 5'-CDP 3Na

Formula: C₉H₁₅N₃O₁₁P₂·H₂O
MW: 469.12 g/mol (anhy)
MDL Number: MFCD00069770
CAS Number: 34393-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a58e7cb7994ca8dcbef1977d65cad3a426a8947&vid=95861625bc7f16969703ab7c3c1d8d7be82fdc71&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR