N-Cyclohexylsulfamic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Cyclohexylsulphamic acid, Cyclohexanesulphamic acid, Cyclamic acid, Cyclohexylsulfamic acid, Cyclohexanesulfamic acid

C0486-500G C0486-25G
TCC0486-500GEA 328.64 CAD
TCC0486-500G TCC0486-025G
N-Cyclohexylsulfamic acid ≥98.0% (by titrimetric analysis)
N-Cyclohexylsulfamic acid

CAS Number: 100-88-9
MDL Number: MFCD00065234
Molecular Formula: C6H13NO3S
Molecular Weight: 179.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 169


Other Product Names / Synonyms: Cyclohexanesulfamic Acid

Formula: C₆H₁₃NO₃S
MW: 179.23 g/mol
Melting Pt: 169 °C
MDL Number: MFCD00065234
CAS Number: 100-88-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0b158dceae3723a10c891982a83a0edbae7cf19&vid=e0b158dceae3723a10c891982a83a0edbae7cf19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR