Allyl cyclohexanepropionate ≥98.0%

Supplier: TCI America

C0485-500ML C0485-25ML
TCC0485-500MLEA 294.2 CAD
TCC0485-500ML TCC0485-025ML
Allyl cyclohexanepropionate ≥98.0%
Allyl cyclohexanepropionate

CAS Number: 2705-87-5 MDL Number: MFCD00059557 Molecular Formula: C12H20O2 Molecular Weight: 196.29 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 91 Flash Point (°C): 126 Specific Gravity (20/20): 0.95


Form: Clear Liquid


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Cyclohexanepropionic Acid Allyl Ester, 3-Cyclohexylpropionic Acid Allyl Ester

Formula: C₁₂H₂₀O₂
Boiling Pt: 91 °C
Density: 0.95 (20 °C)
Flash Pt: 126 °C
MDL Number: MFCD00059557
CAS Number: 2705-87-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9254211a63c907a95b8c349ca8b7616c882e8395&vid=9254211a63c907a95b8c349ca8b7616c882e8395&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR