Trimethyl-trans-aconitate ≥97.0%

Supplier: TCI America

Synonyms: trans-Aconitic Acid Trimethyl Ester

A0725-5G A0725-25G
TCA0725-5GEA 155.31 CAD
TCA0725-5G TCA0725-025G
Trimethyl-trans-aconitate ≥97.0%
Trimethyl-trans-aconitate

CAS Number: 4271-99-2
MDL Number: MFCD00059304
Molecular Formula: C9H12O6
Molecular Weight: 216.19
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Specific Gravity (20/20): 1.22


Other Product Names / Synonyms: trans-Aconitic Acid Trimethyl Ester

Formula: C₉H₁₂O₆
MW: 216.19 g/mol
Boiling Pt: 160 °C
Density: 1.22 (20 °C)
MDL Number: MFCD00059304
CAS Number: 4271-99-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acd3aaef59d51a3f786a856f1b2793576a87a079&vid=acd3aaef59d51a3f786a856f1b2793576a87a079&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR