Diethyl-1,2-cyclohexanedicarboxylate ≥95.0%

Supplier: TCI America

C0476-500G C0476-25G
TCC0476-500GEA 492.96 CAD
TCC0476-500G TCC0476-025G
Diethyl-1,2-cyclohexanedicarboxylate ≥95.0%
Diethyl-1,2-cyclohexanedicarboxylate

CAS Number: 10138-59-7
MDL Number: MFCD00029449
Molecular Formula: C12H20O4
Molecular Weight: 228.29
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: 1,2-Cyclohexanedicarboxylic Acid Diethyl Ester

Formula: C₁₂H₂₀O₄
Boiling Pt: 135 °C
Density: 1.05 (20 °C)
MDL Number: MFCD00029449
CAS Number: 10138-59-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f53efc1be0e2a87e4964c67480426dcb277bb9d&vid=8f53efc1be0e2a87e4964c67480426dcb277bb9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR