trans-1,4-Cyclohexanedicarboxylic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

C0475-500G C0475-25G C0475-100G
TCC0475-500GEA 644.64 CAD
TCC0475-500G TCC0475-025G TCC0475-100G
trans-1,4-Cyclohexanedicarboxylic acid ≥97.0% (by GC, titration analysis)
trans-1,4-Cyclohexanedicarboxylic acid

CAS Number: 619-82-9
MDL Number: MFCD00066203
Molecular Formula: C8H12O4
Molecular Weight: 172.18
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal

Formula: C₈H₁₂O₄
MW: 172.18 g/mol
MDL Number: MFCD00066203
CAS Number: 619-82-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ef28fdbb866cdddf6c53d5762858378305e5130&vid=8ef28fdbb866cdddf6c53d5762858378305e5130&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR