Isopropyl cyclohexanecarboxylate ≥98.0%

Supplier: TCI America

C0472-500ML C0472-25ML
TCC0472-500MLEA 925.56 CAD
TCC0472-500ML TCC0472-025ML
Isopropyl cyclohexanecarboxylate ≥98.0%
Isopropyl cyclohexanecarboxylate

CAS Number: 6553-80-6
MDL Number: MFCD00058949
Molecular Formula: C10H18O2
Molecular Weight: 170.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 131
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: Cyclohexanecarboxylic Acid Isopropyl Ester

Formula: C₁₀H₁₈O₂
Boiling Pt: 131 °C
Density: 0.94 (20 °C)
MDL Number: MFCD00058949
CAS Number: 6553-80-6

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR