N-(2-Cyanoethyl)-N-ethylaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-(Ethyl(phenyl)amino)propanenitrile, 3-(N-Ethyl-N-phenylamino)propionitrile

C0451-500G C0451-25G
TCC0451-500GEA 445.56 CAD
TCC0451-500G TCC0451-025G
N-(2-Cyanoethyl)-N-ethylaniline ≥98.0% (by GC, titration analysis)
N-(2-Cyanoethyl)-N-ethylaniline

CAS Number: 148-87-8
MDL Number: MFCD00019858
Molecular Formula: C11H14N2
Molecular Weight: 174.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 176
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: 3-(N-Ethyl-N-phenylamino)propionitrile

Formula: C₁₁H₁₄N₂
MW: 174.25 g/mol
Boiling Pt: 176 °C
Density: 1.03 (20 °C)
MDL Number: MFCD00019858
CAS Number: 148-87-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9087cfbbab8e2595ed699344643e45559666ac9a&vid=9087cfbbab8e2595ed699344643e45559666ac9a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR