Cyanine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C0436-5G C0436-1G
TCC0436-5GEA 718.9 CAD
TCC0436-5G TCC0436-001G
Cyanine ≥98.0% (by titrimetric analysis)
Cyanine

CAS Number: 523-42-2
MDL Number: MFCD00050330
Molecular Formula: C29H35IN2
Molecular Weight: 538.52
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Cyanine & Squarylium Dyes (Functional Organic Dyes), Bioscience, Stains & Dyes
Other Product Names / Synonyms: Quinoline Blue

Formula: C₂₉H₃₅IN₂
MDL Number: MFCD00050330
CAS Number: 523-42-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f629c742140e0582117aec52c57268f79ca2533&vid=9f629c742140e0582117aec52c57268f79ca2533&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR