4-tert-Butylcyclohexyl acetate (mixture of cis and trans isomers) ≥96.0%

Supplier: TCI America

A0723-500ML A0723-25ML
TCA0723-500MLEA 223.16 CAD
TCA0723-500ML TCA0723-025ML
4-tert-Butylcyclohexyl acetate (mixture of cis and trans isomers) ≥96.0%
4-tert-Butylcyclohexyl acetate (mixture of cis and trans isomers)

CAS Number: 32210-23-4
MDL Number: MFCD00019354
Molecular Formula: C12H22O2
Molecular Weight: 198.31
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 230
Flash Point (°C): 102
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: Acetic Acid 4-tert-Butylcyclohexyl Ester

Formula: C₁₂H₂₂O₂
Boiling Pt: 230 °C
Density: 0.94 (20 °C)
Flash Pt: 102 °C
MDL Number: MFCD00019354
CAS Number: 32210-23-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f8b7a66168706b3747492f2dc86dccbf860c117&vid=742d5b92ad17a88f7bca39730d65a6adc37b39fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR