Butyl crotonate ≥97.0%

Supplier: TCI America

Synonyms: Crotonic Acid Butyl Ester

C0417-25ML C0417-500ML
TCC0417-025MLEA 52.14 CAD
TCC0417-025ML TCC0417-500ML
Butyl crotonate ≥97.0%
Butyl crotonate

CAS Number: 7299-91-4
MDL Number: MFCD00053794
Molecular Formula: C8H14O2
Molecular Weight: 142.20
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 180
Flash Point (°C): 64
Specific Gravity (20/20): 0.90


Other Product Names / Synonyms: Crotonic Acid Butyl Ester

Formula: C₈H₁₄O₂
Boiling Pt: 180 °C
Density: 0.90 (20 °C)
Flash Pt: 64 °C
MDL Number: MFCD00053794
CAS Number: 7299-91-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81b10b4f9b740975ca68f73fdc8a7df121cace6e&vid=81b10b4f9b740975ca68f73fdc8a7df121cace6e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR