Creatine phosphate disodium salt, hydrate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Sodium creatine phosphate dibasic, N-Methyl-N-(N-phosphonocarbamimidoyl)glycine, disodium salt, Sodium creatine phosphate, Phosphocreatine disodium salt

C0397-1G C0397-25G
TCC0397-001GEA 67.72 CAD
TCC0397-001G TCC0397-025G
Creatine phosphate disodium salt, hydrate ≥98.0% (by HPLC)
Creatine phosphate disodium salt

CAS Number: 922-32-7 MDL Number: MFCD00014351 Molecular Formula: C4H10N3O5P Molecular Weight: 255.08 Purity/Analysis Method: 98.0% (HPLC) Form: Crystal Storage Temperature: 0-10°C


Other Product Names / Synonyms: Phosphocreatine Sodium Salt, Creatine Phosphate Sodium Salt, Sodium Phosphocreatine

Formula: C₄H₈N₃O₅P·2Na
MW: 255.077 g/mol
MDL Number: MFCD00014351
CAS Number: 922-32-7
EINECS: 213-074-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05e56276c0712aa81ff56ec20dca788d9d2a2a79&vid=05e56276c0712aa81ff56ec20dca788d9d2a2a79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR