1-Adamantanemethylamine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A0721-1G A0721-5G
TCA0721-001GEA 205.4 CAD
TCA0721-001G TCA0721-5G
1-Adamantanemethylamine ≥98.0% (by titrimetric analysis)
1-Adamantanemethylamine

CAS Number: 17768-41-1
MDL Number: MFCD00074750
Molecular Formula: C11H19N
Molecular Weight: 165.28
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 97
Flash Point (°C): 92
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 1-Aminomethyladamantane

Formula: C₁₁H₁₉N
Boiling Pt: 97 °C
Density: 1.00 (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00074750
CAS Number: 17768-41-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e98a26a9f04206c03d5f9b26c1c299689ef9ab17&vid=e98a26a9f04206c03d5f9b26c1c299689ef9ab17&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR