Copper PAN
Supplier: TCI America
Synonyms:
Bis[1-(2-pyridylazo)-2-naphtholato]copper(II)
C0385-1G
C0385-10G
TCC0385-001GEA
64.46
CAD
TCC0385-001G
TCC0385-010G
Copper PAN
Copper PAN
CAS Number: 23299-85-6
MDL Number: MFCD00067176
Form: Crystal
Color: Yellow Red
Other Product Names / Synonyms: Bis[1-(2-pyridylazo)-2-naphtholato]copper(II)
MDL Number:
MFCD00067176 CAS Number: 23299-85-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...