Cobalt (III) acetylacetonate ≥98.0% for synthesis

Supplier: TCI America

Synonyms: Cobalt (III) tris(2,4-pentanedionato-O,O'), Cobalt (III) 2,4-pentanedionate, Cobalt tris(acetylacetonate), Tris(2,4-pentanedionato-O,O')cobalt (III)

C0374-500G C0374-25G C0374-100G
TCC0374-500GEA 684.14 CAD
TCC0374-500G TCC0374-025G TCC0374-100G
Cobalt (III) acetylacetonate ≥98.0% for synthesis
Cobalt (III) acetylacetonate

CAS Number: 21679-46-9
MDL Number: MFCD00013488
Molecular Formula: C15H21CoO6
Molecular Weight: 356.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Flash Point (°C): 180


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts
Other Product Names / Synonyms: Acetylacetone Cobalt(III) Salt, Cobalt(III) Acetylacetonate

Formula: C₁₅H₂₁CoO₆
MW: 356.26 g/mol
Flash Pt: 180 °C
MDL Number: MFCD00013488
CAS Number: 21679-46-9
EINECS: 244-527-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45f6c2c23cf0b4c8c2361be2d369e0127bb6ac3e&vid=45f6c2c23cf0b4c8c2361be2d369e0127bb6ac3e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR