Cobalt(II) acetylacetonate dihydrate ≥97% (dried basis)
Supplier: TCI America
Synonyms:
Bis(2,4-pentanedionato)cobalt(II) dihydrate, Acetylacetone Cobalt(II) salt dihydrate
CAS Number: 123334-29-2
MDL Number: MFCD00000014
Molecular Formula: C10H14CoO4
Molecular Weight: 257.15
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Oxidation, Materials Chemistry, Magnetic Metal Complexes (Magnetic Materials)
Other Product Names / Synonyms: Acetylacetone Cobalt(II) Salt, Cobalt(II) Acetylacetonate
Formula:
C₁₀H₁₄CoO₄ MW: 293.18 g/mol |
MDL Number:
MFCD00000014 CAS Number: 123334-29-2 |
|
Specification Test Results
Purity (Chelometric Titration) | min. 97.0% (calcd.on anh.substance) |
Water | 10.0 to 14.0 % |
Solubility in hot Chloroform | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...