Dimethyl citraconate ≥99.0%

Supplier: TCI America

C0364-5G C0364-25G
TCC0364-005GEA 135.88 CAD
TCC0364-005G TCC0364-25G
Dimethyl citraconate ≥99.0%
Dimethyl citraconate

CAS Number: 617-54-9
MDL Number: MFCD00059306
Molecular Formula: C7H10O4
Molecular Weight: 158.15
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 211
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: Citraconic Acid Dimethyl Ester

Formula: C₇H₁₀O₄
Boiling Pt: 211 °C
Density: 1.11 (20 °C)
MDL Number: MFCD00059306
CAS Number: 617-54-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b85df80d8deb927de14fc5769fbcf73e40a711b&vid=3b85df80d8deb927de14fc5769fbcf73e40a711b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR