cis,trans-5,9-Cyclododecadiene-cis-1,2-diol ≥98.0%

Supplier: TCI America

Synonyms: cis-1,2-Dihydroxy-cis,trans-5,9-cyclododecadiene

C0355-5G C0355-25G
TCC0355-5GEA 139.04 CAD
TCC0355-5G TCC0355-025G
cis,trans-5,9-Cyclododecadiene-cis-1,2-diol ≥98.0%
cis,trans-5,9-Cyclododecadiene-cis-1,2-diol

CAS Number: 29118-70-5
MDL Number: MFCD00021191
Molecular Formula: C12H20O2
Molecular Weight: 196.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 159


Other Product Names / Synonyms: cis-1,2-Dihydroxy-cis,trans-5,9-cyclododecadiene

Formula: C₁₂H₂₀O₂
MW: 196.29 g/mol
Melting Pt: 159 °C
MDL Number: MFCD00021191
CAS Number: 29118-70-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c41f322d26ab333d5acd14a3d071f874133519a&vid=1c41f322d26ab333d5acd14a3d071f874133519a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR