Chromium(III) acetylacetonate ≥98.0% for synthesis

Supplier: TCI America
Danger

Synonyms: Tris(2,4-pentanedionato-O,O')chromium (III), Chromium (III) 2,4-pentanedionate, Chromium (III) tris(2,4-pentanedionato-O,O')

C0336-25G C0336-100G
TCC0336-025GEA 37.35 CAD
TCC0336-025G TCC0336-100G
Chromium(III) acetylacetonate ≥98.0% for synthesis
Chromium (III) acetylacetonate

CAS Number: 21679-31-2
MDL Number: MFCD00000015
Molecular Formula: C15H21CrO6
Molecular Weight: 349.32
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 216


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Materials Chemistry, Magnetic Metal Complexes (Magnetic Materials)
Other Product Names / Synonyms: Acetylacetone Chromium(III) Salt, Chromium(III) Acetylacetonate

Formula: C₁₅H₂₁CrO₆
MW: 349.32 g/mol
Boiling Pt: 340
Melting Pt: 215.85
Density: 1.266 g/cm³ (20 °C)
MDL Number: MFCD00000015
CAS Number: 21679-31-2
EINECS: 244-526-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b4950f94836339dc93163087b0986591bc07353&vid=9b4950f94836339dc93163087b0986591bc07353&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR