Choline dihydrogen citrate ≥99.0% (by titrimetric analysis)
Supplier: TCI America
C0331-500G
C0331-25G
TCC0331-500GEA
201.77
CAD
TCC0331-500G
TCC0331-025G
Choline dihydrogen citrate ≥99.0% (by titrimetric analysis)
Choline dihydrogen citrate
CAS Number: 77-91-8
MDL Number: MFCD00038734
Molecular Formula: C11H21NO8
Molecular Weight: 295.29
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 107
Formula:
C₁₁H₂₁NO₈ Melting Pt: 107 °C |
MDL Number:
MFCD00038734 CAS Number: 77-91-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...