Tricholine citrate ∼65% in water
Supplier: TCI America
C0330-500G
C0330-25G
TCC0330-500GEA
278.06
CAD
TCC0330-500G
TCC0330-025G
Tricholine citrate ∼65% in water
Tricholine citrate
CAS Number: 546-63-4
MDL Number: MFCD00059985
Form: Clear Liquid
Color: Pale Yellow
Other Product Names / Synonyms: Choline Citrate Solution
MDL Number:
MFCD00059985 CAS Number: 546-63-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...