Choline bromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C0328-25G C0328-500G
TCC0328-025GEA 120.71 CAD
TCC0328-025G TCC0328-500G
Choline bromide ≥98.0% (by titrimetric analysis)
Choline bromide

CAS Number: 1927-06-6
MDL Number: MFCD00054244
Molecular Formula: C5H14BrNO
Molecular Weight: 184.08
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 298


Product Categories: Bioscience, Vitamin Related Compounds
Other Product Names / Synonyms: (2-Hydroxyethyl)trimethylammonium Bromide

Formula: C₅H₁₄BrNO
Melting Pt: 298 °C
MDL Number: MFCD00054244
CAS Number: 1927-06-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7831c78332123fbdc59e945ae3777a6aa05f82ca&vid=7831c78332123fbdc59e945ae3777a6aa05f82ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR