N-Acetyl-DL-histidine monohydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Ac-DL-His-OH·H2O

A0698-5G A0698-1G
TCA0698-5GEA 219.62 CAD
TCA0698-5G TCA0698-001G
N-Acetyl-DL-histidine monohydrate ≥98.0% (by HPLC, titration analysis)
N-Acetyl-DL-histidine monohydrate

CAS Number: 213178-97-3
MDL Number: MFCD00065961
Molecular Formula: C8H11N3O3
Molecular Weight: 197.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 152


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Ac-DL-His-OH

Formula: C₈H₁₃N₃O₄
MW: 215.2 g/mol
Melting Pt: 152 °C
MDL Number: MFCD00065961
CAS Number: 213178-97-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8521587a111e3d195feef8a70ae3b15bd238bc1a&vid=31e71d94528a8d495ba58cbf5fac66babd4fe142&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR