2-Chloro-4'-phenylacetophenone ≥99.0% (by GC, titration analysis)

Supplier: TCI America

C0252-25G C0252-5G
TCC0252-025GEA 279.66 CAD
TCC0252-025G TCC0252-5G
2-Chloro-4'-phenylacetophenone ≥99.0% (by GC, titration analysis)
2-Chloro-4'-phenylacetophenone

CAS Number: 635-84-7
MDL Number: MFCD00058937
Molecular Formula: C14H11ClO
Molecular Weight: 230.69
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 126


Other Product Names / Synonyms: 4-Chloroacetylbiphenyl, 4-Phenylphenacyl Chloride

Formula: C₁₄H₁₁ClO
Melting Pt: 126 °C
MDL Number: MFCD00058937
CAS Number: 635-84-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb46687f847863cb75dbdf4f55d283e93b4f7faa&vid=eb46687f847863cb75dbdf4f55d283e93b4f7faa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR