Chlorophenol red sodium salt
Supplier: TCI America
C0247-5G
C0247-1G
TCC0247-5GEA
94.8
CAD
TCC0247-5G
TCC0247-001G
Chlorophenol red sodium salt
Chlorophenol red sodium salt
CAS Number: 123333-64-2
MDL Number: MFCD00151199
Molecular Formula: C19H12Cl2O5S
Molecular Weight: 445.24
Form: Crystal
Color: Deep Yellow Red
Formula:
C₁₉H₁₂Cl₂O₅S MW: 445.24 g/mol |
MDL Number:
MFCD00151199 CAS Number: 123333-64-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...