Carboxymethoxylamine hemihydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Aminooxyacetic acid hemihydrochloride, O-(Carboxymethyl)hydroxylamine Hemihydrochloride, O-(Carboxymethyl)hydroxylamine hemihydrochloride, (Aminooxy)acetic Acid Hemihydrochloride
CAS Number: 2921-14-4
MDL Number: MFCD00012955
Molecular Formula: C2H5NO3
Molecular Weight: 109.30
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Product Categories: Analytical Science, Derivatization Reagents for MS
Other Product Names / Synonyms: (Aminooxy)acetic Acid Hemihydrochloride, O-(Carboxymethyl)hydroxylamine Hemihydrochloride
Formula:
C₄H₁₁ClN₂O₆ MW: 218.59 g/mol |
MDL Number:
MFCD00012955 CAS Number: 2921-14-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...