4-Chloro-α-methylbenzyl alcohol ≥97.0%

Supplier: TCI America

Synonyms: 1-(4-Chlorophenyl)ethanol, 4-Chlorophenyl methyl carbinol, p-Chloro-α-methylbenzyl alcohol

C0198-10G C0198-25G
TCC0198-010GEA 96.38 CAD
TCC0198-010G TCC0198-025G
4-Chloro-α-methylbenzyl alcohol ≥97.0%
4-Chloro-α-methylbenzyl alcohol

CAS Number: 3391-10-4
MDL Number: MFCD00041038
Molecular Formula: C8H9ClO
Molecular Weight: 156.61
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 136
Flash Point (°C): 113
Specific Gravity (20/20): 1.17


Other Product Names / Synonyms: 1-(4-Chlorophenyl)ethanol, 1-(4-Chlorophenyl)ethyl Alcohol

Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 136 °C
Density: 1.17 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00041038
CAS Number: 3391-10-4
EINECS: 222-223-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b58fb249b9d88d05377db3ae2ee4e0a9187c7349&vid=b58fb249b9d88d05377db3ae2ee4e0a9187c7349&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR