(±)-4-Chloromandelic acid ≥98.0%

Supplier: TCI America

Synonyms: 4-Chloromandelic acid, (±)-4-Chlorophenylglycolic acid

C0193-5G C0193-25G
TCC0193-005GEA 92.59 CAD
TCC0193-005G TCC0193-025G
(±)-4-Chloromandelic acid ≥98.0%
(±)-4-Chloromandelic acid

CAS Number: 492-86-4
MDL Number: MFCD00042724
Molecular Formula: C8H7ClO3
Molecular Weight: 186.59
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 120

Formula: C₈H₇ClO₃
MW: 186.59 g/mol
Boiling Pt: 361
Melting Pt: 120 °C
MDL Number: MFCD00042724
CAS Number: 492-86-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b8291cffba8e781b7d275ef01d111eaefe51815&vid=1b8291cffba8e781b7d275ef01d111eaefe51815&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR