Chlorogenic acid hydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-(3,4-Dihydroxycinnamoyl)quinic acid hydrate, (1S,3R,4R,5R)-3-(((3-(3,4-dihydroxyphenyl)acryloyl)oxy)-1,4,5-trihydroxycyclohexanecarboxylic acid hydrate, 3-O-Caffeoylquinic acid hydrate, (1S,3R,4R,5R)-3-[[3-(3,4-Dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,4,5-trihydroxycyclohexanecarboxylic acid hydrate

C0181-1G C0181-5G
TCC0181-001GEA 136.21 CAD
TCC0181-001G TCC0181-005G
Chlorogenic acid hydrate ≥98.0% (by HPLC, titration analysis)
Chlorogenic acid hydrate

CAS Number: 327-97-9
MDL Number: MFCD00003862
Molecular Formula: C16H18O9
Molecular Weight: 354.31
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 208
Specific rotation [a]20/D: -37 deg (C=1, H2O)


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Crude Medicine Ingredients for Research and Experimental Use, Antioxidants, Phenylpropanoids, Phenylpropanoids (Phytochemicals), Epigenetics Research Reagents
Other Product Names / Synonyms: 3-(3,4-Dihydroxycinnamoyl)quinic Acid

Formula: C₁₆H₁₈O₉
Melting Pt: 208 °C
MDL Number: MFCD00003862
CAS Number: 327-97-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=beb41249a7ecc5635248f61e64830e9f4be29dfd&vid=d4c9f72e12e7c89ab87e0887ce5daeda5e541166&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR