Sodium-4-chlorobenzenesulfonate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C0127-500G C0127-25G
TCC0127-500GEA 1257.68 CAD
TCC0127-500G TCC0127-025G
Sodium-4-chlorobenzenesulfonate ≥98.0% (by HPLC, titration analysis)
Sodium-4-chlorobenzenesulfonate

CAS Number: 5138-90-9
MDL Number: MFCD00014731
Molecular Formula: C6H5ClO3S
Molecular Weight: 214.59
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 4-Chlorobenzenesulfonic Acid Sodium Salt

Formula: C₆H₅ClO₃S
MDL Number: MFCD00014731
CAS Number: 5138-90-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab6c58f392496ff423e223c0fee30bb2e69750f0&vid=ab6c58f392496ff423e223c0fee30bb2e69750f0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR