4-Chloroaniline-3-sulfonic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C0116-500G C0116-25G
TCC0116-500GEA 345.39 CAD
TCC0116-500G TCC0116-025G
4-Chloroaniline-3-sulfonic acid ≥98.0% (by HPLC, titration analysis)
4-Chloroaniline-3-sulfonic acid

CAS Number: 88-43-7
MDL Number: MFCD00025296
Molecular Formula: C6H6ClNO3S
Molecular Weight: 207.63
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 5-Amino-2-chlorobenzenesulfonic Acid

Formula: C₆H₆ClNO₃S
MDL Number: MFCD00025296
CAS Number: 88-43-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9264c7d2797dde5cadd7937a084b78a9a292b3e5&vid=9264c7d2797dde5cadd7937a084b78a9a292b3e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR