N-Chloroacetyl-L-valine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C0107-1G C0107-100MG
TCC0107-001GEA 64.27 CAD
TCC0107-001G TCC0107-100MG
N-Chloroacetyl-L-valine ≥98.0% (by titrimetric analysis)
N-Chloroacetyl-L-valine

CAS Number: 2279-16-5
MDL Number: MFCD00045273
Molecular Formula: C7H12ClNO3
Molecular Weight: 193.63
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 115

Formula: C₇H₁₂ClNO₃
Melting Pt: 115 °C
MDL Number: MFCD00045273
CAS Number: 2279-16-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bb048758a94c495b2ef87c96abf3a45e5a6c7ea&vid=8bb048758a94c495b2ef87c96abf3a45e5a6c7ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR