Chloroacetylglycylglycine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C0099-25G C0099-1G
TCC0099-025GEA 371.3 CAD
TCC0099-025G TCC0099-001G
Chloroacetylglycylglycine ≥98.0% (by titrimetric analysis)
Chloroacetylglycylglycine

CAS Number: 15474-96-1
MDL Number: MFCD00055928
Molecular Formula: C6H9ClN2O4
Molecular Weight: 208.60
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 179

Formula: C₆H₉ClN₂O₄
MW: 208.60 g/mol
Melting Pt: 179 °C
MDL Number: MFCD00055928
CAS Number: 15474-96-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f02050d9e1dc905fb197ee615447dc9c59e3f28&vid=1f02050d9e1dc905fb197ee615447dc9c59e3f28&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR