Chloranilic acid sodium salt hydrate ≥95.0%
Supplier: TCI America
Synonyms:
Sodium Chloranilate, Sodium chloranilate trihydrate
C0081-5G
C0081-25G
TCC0081-005GEA
45.82
CAD
TCC0081-005G
TCC0081-025G
Chloranilic acid sodium salt hydrate ≥95.0%
Chloranilic acid sodium salt hydrate
CAS Number: 36275-66-8
MDL Number: MFCD00013787
Molecular Formula: C6H2Cl2O4
Molecular Weight: 252.94
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Other Product Names / Synonyms: Sodium Chloranilate
Formula:
C₆H₂Cl₂O₄ MW: 252.94 g/mol |
MDL Number:
MFCD00013787 CAS Number: 36275-66-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...